Stampa…

trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: trans-4'-(4-Methylphenyl)[1,1'-bi(cyclohexyl)]-4-one, (1'r,4'r)-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one, trans-4'-(p-tolyl)-[1,1'-Bi(cyclohexan)]-4-one

T3465-1G T3465-5G
TCIAT3465-1GEA 27 EUR
TCIAT3465-1G TCIAT3465-5G
trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one ≥98.0% (by GC)
trans-4'-(p-Tolyl)-[1,1'-bi(cyclohexan)]-4-one
Formula: C₁₉H₂₆O
Peso molecolare: 270,42 g/mol
MDL Number: MFCD13194928
CAS Number: 125962-80-3
EINECS: 435-110-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR