Stampa…

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

T3441-1G T3441-5G
TCIAT3441-1GEA 96 EUR
TCIAT3441-1G TCIAT3441-5G
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Formula: C₁₈H₂₀BNO₂
MDL Number: MFCD07784368
CAS Number: 1242412-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6c65c05173d63cbcd37d46f5cf5743e4a2e2d0c&vid=d6c65c05173d63cbcd37d46f5cf5743e4a2e2d0c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 195.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR