4,4',4''-Tris[9,9-dimethylfluoren-2-yl(phenyl)amino]triphenylamine ≥98.0% (by HPLC)
Fornitore: TCI
Formula:
C₈₁H₆₆N₄ |
CAS Number:
303111-06-0 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...