Stampa…

2,2'':7'',2''''-Ter-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: TSBF

T3433-1G T3433-200MG
TCIAT3433-1GEA 353 EUR
TCIAT3433-1G TCIAT3433-200MG
2,2'':7'',2''''-Ter-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)
2,2'':7'',2''''-Ter-9,9'-spirobi[9H-fluorene]
Formula: C₇₅H₄₄
Peso molecolare: 945,18 g/mol
Punto di fusione: 457°C
MDL Number: MFCD12022449
CAS Number: 518997-91-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 93.00 to 95.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR