2,2,2-Trifluoro-3'-nitroacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3'-Nitro-2,2,2-trifluoroacetophenone, 2,2,2-Trifluoro-1-(3-nitrophenyl)ethanone
T3426-1G
T3426-5G
TCIAT3426-1GEA
124
EUR
TCIAT3426-1G
TCIAT3426-5G
2,2,2-Trifluoro-3'-nitroacetophenone ≥98.0% (by GC)
2,2,2-Trifluoro-3'-nitroacetophenone
Formula:
C₈H₄F₃NO₃ Peso molecolare: 219,12 g/mol Punto di ebollizione: 113°C |
MDL Number:
MFCD00034541 CAS Number: 657-15-8 EINECS: 211-516-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...