Stampa…

Tetrakis(4-bromophenyl)ethylene ≥97.0% (by GC)

Fornitore: TCI
Attenzione

T3423-1G T3423-5G
TCIAT3423-1GEA 94 EUR
TCIAT3423-1G TCIAT3423-5G
Tetrakis(4-bromophenyl)ethylene ≥97.0% (by GC)
Tetrakis(4-bromophenyl)ethylene
Formula: C₂₆H₁₆Br₄
MDL Number: MFCD00667761
CAS Number: 61326-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8f3c87801b56e553cb789bd6cc0a6884952d3e4&vid=f8f3c87801b56e553cb789bd6cc0a6884952d3e4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR