1-(2-Tetrahydropyranyl)-1H-pyrazole ≥98.0% (by GC)
Fornitore: TCI
T3408-1G
T3408-5G
TCIAT3408-1GEA
33
EUR
TCIAT3408-1G
TCIAT3408-5G
1-(2-Tetrahydropyranyl)-1H-pyrazole ≥98.0% (by GC)
1-(2-Tetrahydropyranyl)-1H-pyrazole
Formula:
C₈H₁₂N₂O Punto di ebollizione: 65°C Flash Pt: 104°C |
MDL Number:
MFCD11044692 CAS Number: 449758-17-2 |
|
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |