Stampa…

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(4-pyridyl)phenyl]-1,3,2-dioxaborolane, 3-(4-Pyridyl)phenylboronic acid Pinacol ester, 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyridine

T3399-200MG T3399-1G
TCIAT3399-200MGEA 71 EUR
TCIAT3399-200MG TCIAT3399-1G
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Formula: C₁₇H₂₀BNO₂
Peso molecolare: 281,16 g/mol
MDL Number: MFCD20923942
CAS Number: 1009033-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9900e8a2610f27d1eebcf2dba34345e3a6e2b6eb&vid=9900e8a2610f27d1eebcf2dba34345e3a6e2b6eb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR