Stampa…

2-(Bromomethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(2-(bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Bromomethyl)phenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide

T3393-1G T3393-5G
TCIAT3393-1GEA 148 EUR
TCIAT3393-1G TCIAT3393-5G
2-(Bromomethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-(Bromomethyl)benzeneboronic acid pinacol ester
Formula: C₁₃H₁₈BBrO₂
Peso molecolare: 297,00 g/mol
MDL Number: MFCD01632210
CAS Number: 377780-72-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c7a5a87ae0b0d383ffee26219febcaa15d96317&vid=7c7a5a87ae0b0d383ffee26219febcaa15d96317&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR