Stampa…

2,4,6-Tribromo-3-hydroxybenzaldehyde ≥98.0% (by GC)

Fornitore: TCI

T3392-25G T3392-5G
TCIAT3392-25GEA 226 EUR
TCIAT3392-25G TCIAT3392-5G
2,4,6-Tribromo-3-hydroxybenzaldehyde ≥98.0% (by GC)
2,4,6-Tribromo-3-hydroxybenzaldehyde
Formula: C₇H₃Br₃O₂
MDL Number: MFCD00017320
CAS Number: 2737-22-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR