Stampa…

3-(Trifluoromethoxy)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Trifluoromethoxy)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethoxy)benzene

T3377-1G T3377-5G
TCIAT3377-1GEA 42 EUR
TCIAT3377-1G TCIAT3377-5G
3-(Trifluoromethoxy)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Trifluoromethoxy)phenylboronic acid pinacol ester
Formula: C₁₃H₁₆BF₃O₃
Peso molecolare: 288,0706 g/mol
MDL Number: MFCD05863919
CAS Number: 262376-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50aeac4ba41769109c007a717093d22fb9b01df&vid=c50aeac4ba41769109c007a717093d22fb9b01df&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR