Stampa…

1,3,5-Tri(p-tolyl)benzene ≥96.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,3,5-Tris(4-Methylphenyl)benzene, 4,4''-Dimethyl-5'-(p-tolyl)-1,1':3',1''-terphenyl

T3370-1G T3370-5G
TCIAT3370-1GEA 60 EUR
TCIAT3370-1G TCIAT3370-5G
1,3,5-Tri(p-tolyl)benzene ≥96.0% (by GC)
1,3,5-Tri(p-tolyl)benzene
Formula: C₂₇H₂₄
Peso molecolare: 348,49 g/mol
MDL Number: MFCD00192798
CAS Number: 50446-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a1131a642c6e91364ad8165ae88412591e728c9&vid=1a1131a642c6e91364ad8165ae88412591e728c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR