N,N,N'-Triphenyl-1,4-phenylenediamine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
T3367-200MG
T3367-1G
TCIAT3367-200MGEA
55
EUR
TCIAT3367-200MG
TCIAT3367-1G
N,N,N'-Triphenyl-1,4-phenylenediamine ≥98.0% (by GC)
N,N,N'-Triphenyl-1,4-phenylenediamine
Formula:
C₂₄H₂₀N₂ |
MDL Number:
MFCD27939629 CAS Number: 19606-98-5 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...