Stampa…

1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane ≥97.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

T3356-1G T3356-5G
TCIAT3356-1GEA 105 EUR
TCIAT3356-1G TCIAT3356-5G
1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane ≥97.0% (by HPLC, total nitrogen)
1,4,7,10-Tetrabenzyl-1,4,7,10-tetraazacyclododecane
Formula: C₃₆H₄₄N₄
Peso molecolare: 532,78 g/mol
MDL Number: MFCD05864401
CAS Number: 18084-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f806865bc8249b179283cffcc697ea60df7838d&vid=0f806865bc8249b179283cffcc697ea60df7838d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 143.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR