Stampa…

Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphite, Phosphorous acid tris(1,1,1,3,3,3-hexafluoro-2-propyl) ester

T3353-1G T3353-5G
TCIAT3353-1GEA 146 EUR
TCIAT3353-1G TCIAT3353-5G
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite ≥98.0% (by GC)
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite
Formula: C₉H₃F₁₈O₃P
Peso molecolare: 532,07 g/mol
Punto di ebollizione: 130°C
MDL Number: MFCD00192561
CAS Number: 66470-81-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR