Stampa…

Trimethoxy(pentafluorophenyl)silane ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Trimethoxy(perfluorophenyl)silane

T3352-1G T3352-5G
TCIAT3352-1GEA 81 EUR
TCIAT3352-1G TCIAT3352-5G
Trimethoxy(pentafluorophenyl)silane ≥97.0% (by GC)
Trimethoxy(pentafluorophenyl)silane
Formula: C₉H₉F₅O₃Si
Peso molecolare: 288.25 g/mol
MDL Number: MFCD04039205
CAS Number: 223668-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f98d57ebad8019a8db82ea7ef097c88a46d6663&vid=2f98d57ebad8019a8db82ea7ef097c88a46d6663&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR