Stampa…

4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(4-Trifluoromethoxyphenoxy)piperidine

T3349-1G T3349-5G
TCIAT3349-1GEA 40 EUR
TCIAT3349-1G TCIAT3349-5G
4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97.0% (by GC, titration analysis)
4-[4-(Trifluoromethoxy)phenoxy]piperidine
Formula: C₁₂H₁₄F₃NO₂
Peso molecolare: 261,25 g/mol
MDL Number: MFCD06656166
CAS Number: 287952-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=210ecd52240b9370258d21bb5262e231e244a5e1&vid=210ecd52240b9370258d21bb5262e231e244a5e1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 74.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR