2,3-Thiophenedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Thieno[2,3-c]furan-4,6-dione
T3342-1G
T3342-5G
TCIAT3342-1GEA
126
EUR
TCIAT3342-1G
TCIAT3342-5G
2,3-Thiophenedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
2,3-Thiophenedicarboxylic anhydride
Formula:
C₆H₂O₃S Peso molecolare: 154,14 g/mol |
MDL Number:
MFCD09832453 CAS Number: 6007-83-6 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...