Tris(4-biphenylyl)amine ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
Tri([1,1'-biphenyl]-4-yl)amine
T3327-1G
T3327-5G
TCIAT3327-1GEA
118
EUR
TCIAT3327-1G
TCIAT3327-5G
Tris(4-biphenylyl)amine ≥98.0% (by HPLC)
Tris(4-biphenylyl)amine
Formula:
C₃₆H₂₇N Peso molecolare: 473,62 g/mol |
MDL Number:
MFCD30470498 CAS Number: 6543-20-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...