N-Methoxy-N-methyltrifluoroacetamide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,2,2-Trifluoro-N-methoxy-N-methylacetamide
T3326-5G
T3326-25G
TCIAT3326-5GEA
67
EUR
TCIAT3326-5G
TCIAT3326-25G
N-Methoxy-N-methyltrifluoroacetamide ≥98.0% (by GC)
N-Methoxy-N-methyltrifluoroacetamide
Formula:
C₄H₆F₃NO₂ Peso molecolare: 157,09 g/mol Punto di ebollizione: 52°C Flash Pt: 47°C |
MDL Number:
MFCD00077565 CAS Number: 104863-67-4 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...