Stampa…
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms: 2',3,4,5-Tetrafluoro-4''-propyl-p-terphenyl
T3318-1G T3318-5G
TCIAT3318-1GEA 57 EUR
TCIAT3318-1G TCIAT3318-5G
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
2',3,4,5-Tetrafluoro-4''-propyl-1,1':4',1''-terphenyl
Formula: C₂₁H₁₆F₄
Peso molecolare: 344,35 g/mol
MDL Number: MFCD12964272
CAS Number: 205806-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6f297e6c65db10554b0e76307ffea140d85d404&vid=f6f297e6c65db10554b0e76307ffea140d85d404&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 61.0 to 67.0 deg-C