Stampa…

N,N,N',N'-Tetraphenyl-1,4-phenylenediamine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,4-Bis(diphenylamino)benzene

T3309-1G T3309-5G
TCIAT3309-1GEA 36 EUR
TCIAT3309-1G TCIAT3309-5G
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine ≥98.0% (by GC)
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine
Formula: C₃₀H₂₄N₂
Peso molecolare: 412,54 g/mol
MDL Number: MFCD08276846
CAS Number: 14118-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b26743c58ae82cc63f0c64a618eb81392ff5be40&vid=b26743c58ae82cc63f0c64a618eb81392ff5be40&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 201.0 to 205.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR