Stampa…

Trihexyphenidyl hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: alpha-Cyclohexyl-alpha-phenyl-1-piperidinepropanol hydrochloride, Benzhexol hydrochloride, 1-Cyclohexyl-1-phenyl-3-(1-piperidyl)-1-propanol hydrochloride

T3303-5G T3303-25G
TCIAT3303-5GEA 91 EUR
TCIAT3303-5G TCIAT3303-25G
Trihexyphenidyl hydrochloride ≥98.0% (by HPLC, titration analysis)
Trihexyphenidyl hydrochloride
Formula: C₂₀H₃₁NO·HCl
Peso molecolare: 337,93 g/mol
MDL Number: MFCD00058212
CAS Number: 52-49-3
Merck Index: 14,09695
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b219cb5e75fbe2cffde63b46f8a3e9b48194012c&vid=b219cb5e75fbe2cffde63b46f8a3e9b48194012c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR