Stampa…

2-(2,2,2-Trifluoroethoxy)aniline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

T3284-1G T3284-5G
TCIAT3284-1GEA 46 EUR
TCIAT3284-1G TCIAT3284-5G
2-(2,2,2-Trifluoroethoxy)aniline ≥98.0% (by GC, titration analysis)
2-(2,2,2-Trifluoroethoxy)aniline
Formula: C₈H₈F₃NO
Peso molecolare: 191,15 g/mol
MDL Number: MFCD00077605
CAS Number: 57946-60-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR