Stampa…

4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4,4,4-Trifluoro-1-(4-methoxyphenyl)-1,3-butanedione

T3283-1G T3283-5G
TCIAT3283-1GEA 66 EUR
TCIAT3283-1G TCIAT3283-5G
4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione ≥98.0% (by GC)
4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione
Formula: C₁₁H₉F₃O₃
Peso molecolare: 246,19 g/mol
MDL Number: MFCD03238723
CAS Number: 15191-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa57e0d0c6b4f7efb90d806c39df100d344af736&vid=fa57e0d0c6b4f7efb90d806c39df100d344af736&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR