Triisopropyl[(trimethylsilyl)ethynyl]silane ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
T3271-1ML
T3271-5ML
TCIAT3271-1MLEA
63
EUR
TCIAT3271-1ML
TCIAT3271-5ML
Triisopropyl[(trimethylsilyl)ethynyl]silane ≥97.0% (by GC)
Triisopropyl[(trimethylsilyl)ethynyl]silane
Formula:
C₁₄H₃₀Si₂ Punto di ebollizione: 57°C |
MDL Number:
MFCD28975120 CAS Number: 107474-02-2 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...