Stampa…

2-(Trifluoromethyl)isonicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(Trifluoromethyl)pyridine-4-carboxylic acid

T3269-200MG T3269-1G
TCIAT3269-200MGEA 30 EUR
TCIAT3269-200MG TCIAT3269-1G
2-(Trifluoromethyl)isonicotinic acid ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)isonicotinic acid
Formula: C₇H₄F₃NO₂
Peso molecolare: 191,11 g/mol
MDL Number: MFCD07774132
CAS Number: 131747-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56190799bb1e2b0aed911d74fc6b9f00c510c0d0&vid=56190799bb1e2b0aed911d74fc6b9f00c510c0d0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point gt. 219.0 and le 223.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR