Stampa…

trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: trans-3,4,5-Trifluoro-4'-(4-propylcyclohexyl)biphenyl, 3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)biphenyl, 3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

T3260-1G T3260-5G
TCIAT3260-1GEA 32 EUR
TCIAT3260-1G TCIAT3260-5G
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl ≥98.0% (by GC)
trans-3,4,5-Trifluoro-4'-(4-n-propylcyclohexyl)biphenyl
Formula: C₂₁H₂₃F₃
Peso molecolare: 332,41 g/mol
MDL Number: MFCD09839001
CAS Number: 132123-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba17c364f482d7d1f72edc8d3b382ca38ed0852&vid=3ba17c364f482d7d1f72edc8d3b382ca38ed0852&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR