Tris(2,2'-bipyridine)cobalt(III) tris(hexafluorophosphate) ≥93.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cobalt(3+), tris(2,2′-bipyridine-κN1,κN1′)-, (OC-6-11)-, hexafluorophosphate(1-) (1:3)
T3256-200MG
T3256-1G
TCIAT3256-200MGEA
122
EUR
TCIAT3256-200MG
TCIAT3256-1G
Tris(2,2'-bipyridine)cobalt(III) tris(hexafluorophosphate) ≥93.0% (by titrimetric analysis)
Tris(2,2'-bipyridine)cobalt(III) tris(hexafluorophosphate)
Formula:
C₃₀H₂₄CoF₁₈N₆P₃ Peso molecolare: 962,39 g/mol |
MDL Number:
MFCD28976173 CAS Number: 28277-53-4 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Chelometric Titration) | min. 93.0 % |
Elemental analysis (Nitrogen) | 8.10 to 9.30 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...