Stampa…

2,2':6',2''-Terpyridine-4'-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: [2,2':6',2''-Terpyridine]-4'-carboxylic acid

T3245-200MG T3245-1G
TCIAT3245-200MGEA 54 EUR
TCIAT3245-200MG TCIAT3245-1G
2,2':6',2''-Terpyridine-4'-carboxylic acid ≥98.0% (by GC, titration analysis)
2,2':6',2''-Terpyridine-4'-carboxylic acid
Formula: C₁₆H₁₁N₃O₂
Peso molecolare: 277,283 g/mol
MDL Number: MFCD04114290
CAS Number: 148332-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14c8c5da710860824767a9e6cf3918c028e57a83&vid=14c8c5da710860824767a9e6cf3918c028e57a83&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR