Stampa…
Thieno[3,2-b]thiophene-2-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 2-Cyanothieno[3,2-b]thiophene
T3221-200MG T3221-1G
TCIAT3221-200MGEA 75 EUR
TCIAT3221-200MG TCIAT3221-1G
Thieno[3,2-b]thiophene-2-carbonitrile ≥98.0% (by GC)
Thieno[3,2-b]thiophene-2-carbonitrile
Formula: C₇H₃NS₂
Peso molecolare: 165,23 g/mol
MDL Number: MFCD08059461
CAS Number: 40985-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748842ad6d5a2b79aae30798dc22a7eec8ca0e8a&vid=748842ad6d5a2b79aae30798dc22a7eec8ca0e8a&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C