Stampa…

1,3,5-Tris(3,5-dibromophenyl)benzene ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3,3'',5,5''-Tetrabromo-5'-(3,5-dibromophenyl)-1,1':3',1''-terphenyl

T3213-200MG T3213-1G
TCIAT3213-200MGEA 84 EUR
TCIAT3213-200MG TCIAT3213-1G
1,3,5-Tris(3,5-dibromophenyl)benzene ≥95.0% (by HPLC)
1,3,5-Tris(3,5-dibromophenyl)benzene
Formula: C₂₄H₁₂Br₆
Peso molecolare: 779,78 g/mol
Punto di fusione: 315°C
MDL Number: MFCD13193235
CAS Number: 29102-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa3f7bf0a9fbbadad4cdecd98d6a140348228034&vid=fa3f7bf0a9fbbadad4cdecd98d6a140348228034&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR