Stampa…

Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tris(1,10-phenanthroline)ruthenium(II) Bis(hexafluorophosphate)

T3208-200MG T3208-1G
TCIAT3208-200MGEA 50 EUR
TCIAT3208-200MG TCIAT3208-1G
Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) ≥98.0% (by HPLC)
Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate)
Formula: C₃₆H₂₄F₁₂N₆P₂Ru
Peso molecolare: 931,63 g/mol
MDL Number: MFCD28369263
CAS Number: 60804-75-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=160fd76246b1cf9513a6392fba8cc6c7f69eb763&vid=160fd76246b1cf9513a6392fba8cc6c7f69eb763&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.50 to 9.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR