Stampa…

2,3,5,6-Tetrabromo-p-xylene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2,4,5-Tetrabromo-3,6-dimethylbenzene

T3202-1G T3202-5G
TCIAT3202-1GEA 37 EUR
TCIAT3202-1G TCIAT3202-5G
2,3,5,6-Tetrabromo-p-xylene ≥98.0% (by GC)
2,3,5,6-Tetrabromo-p-xylene
Formula: C₈H₆Br₄
Peso molecolare: 421,75 g/mol
MDL Number: MFCD00010353
CAS Number: 23488-38-2
EINECS: 245-688-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d335ee0573bc8061ffc662f7e6d538cd1884c19c&vid=d335ee0573bc8061ffc662f7e6d538cd1884c19c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR