Stampa…

4-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methoxyphenylboronic acid Pinacol ester

T3196-5G T3196-25G
TCIAT3196-5GEA 51 EUR
TCIAT3196-5G TCIAT3196-25G
4-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₃
Peso molecolare: 234.1 g/mol
MDL Number: MFCD05663864
CAS Number: 171364-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fef0761aed5029dc526c40c2139aed52e713bcf3&vid=fef0761aed5029dc526c40c2139aed52e713bcf3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR