Stampa…

2-Methylphenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4,5,5-Tetramethyl-2-(o-tolyl)-1,3,2-dioxaborolane, 2-Methylphenylboronic acid, pinacol ester, o-Tolylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene

T3195-1G T3195-5G
TCIAT3195-1GEA 38 EUR
TCIAT3195-1G TCIAT3195-5G
2-Methylphenylboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
2-Methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₂
Peso molecolare: 218,1 g/mol
MDL Number: MFCD06795658
CAS Number: 195062-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa505edd3639901a83bdf95663452841d264fb6&vid=6aa505edd3639901a83bdf95663452841d264fb6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR