Stampa…

3-(Trimethylsilylethynyl)thiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Trimethyl(thiophen-3-ylethynyl)silane

T3190-1G T3190-5G
TCIAT3190-1GEA 35 EUR
TCIAT3190-1G TCIAT3190-5G
3-(Trimethylsilylethynyl)thiophene ≥98.0% (by GC)
3-(Trimethylsilylethynyl)thiophene
Formula: C₉H₁₂SSi
Peso molecolare: 180,34 g/mol
MDL Number: MFCD04039959
CAS Number: 130995-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45320057e477a64c0cfc6504ce5a920bde0ec868&vid=45320057e477a64c0cfc6504ce5a920bde0ec868&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR