Tetradecylphosphonic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
T3183-1G
T3183-5G
TCIAT3183-1GEA
55
EUR
TCIAT3183-1G
TCIAT3183-5G
Tetradecylphosphonic acid ≥98.0% (by titrimetric analysis)
Tetradecylphosphonic acid
Formula:
C₁₄H₃₁O₃P |
MDL Number:
MFCD00015834 CAS Number: 4671-75-4 EINECS: 225-118-1 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...