Stampa…

(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

T3174-1G T3174-5G
TCIAT3174-1GEA 53 EUR
TCIAT3174-1G TCIAT3174-5G
(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) ≥98.0% (by HPLC)
(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers)
Formula: C₈H₄F₃N₃O
Peso molecolare: 215,14 g/mol
MDL Number: MFCD00593044
CAS Number: 183266-61-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09fd246b736c2fa472d30133c7c4b640d5b344d6&vid=09fd246b736c2fa472d30133c7c4b640d5b344d6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR