Stampa…

3-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

T3162-1G T3162-5G
TCIAT3162-1GEA 17 EUR
TCIAT3162-1G TCIAT3162-5G
3-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
Peso molecolare: 248,083 g/mol
MDL Number: MFCD03411930
CAS Number: 269409-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=626fe6b268ff01238a59d352db0e47e6fe0f2839&vid=626fe6b268ff01238a59d352db0e47e6fe0f2839&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR