Stampa…

2-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Formylphenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Formylphenylboronic acid pinacol ester, 2-(2-Formylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3161-1G T3161-5G
TCIAT3161-1GEA 34 EUR
TCIAT3161-1G TCIAT3161-5G
2-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC, titration analysis)
2-Formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₇BO₃
Peso molecolare: 232,08 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD07363841
CAS Number: 380151-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85c1bd6a0a5f0e0f7ea10702390ae3c2a92455cc&vid=85c1bd6a0a5f0e0f7ea10702390ae3c2a92455cc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR