Stampa…

8-(Trifluoromethyl)-4-quinolinol ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-Hydroxy-8-(trifluoromethyl)quinoline

T3160-1G T3160-5G
TCIAT3160-1GEA 58 EUR
TCIAT3160-1G TCIAT3160-5G
8-(Trifluoromethyl)-4-quinolinol ≥98.0% (by GC, titration analysis)
8-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
MDL Number: MFCD00134577
CAS Number: 23779-96-6
EINECS: 245-879-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=689d9fab2028be25a94ebaf7ec184c968359fdfd&vid=689d9fab2028be25a94ebaf7ec184c968359fdfd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR