Stampa…

a-Tosylbenzyl isocyanide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide

T3157-1G T3157-5G
TCIAT3157-1GEA 314 EUR
TCIAT3157-1G TCIAT3157-5G
a-Tosylbenzyl isocyanide ≥98.0% (by HPLC, total nitrogen)
a-Tosylbenzyl isocyanide
Formula: C₁₅H₁₃NO₂S
Peso molecolare: 271,3342 g/mol
MDL Number: MFCD04114776
CAS Number: 36635-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc8f0da546938a25915db827f36aeb273c6b9363&vid=cc8f0da546938a25915db827f36aeb273c6b9363&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR