3-(Trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(Trifluoromethyl)pyridine-2-carboxylic acid, 3-(Trifluoromethy)pyridine-2-carboxylic acid
T3155-100MG
T3155-500MG
TCIAT3155-100MGEA
27
EUR
TCIAT3155-100MG
TCIAT3155-500MG
3-(Trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)
3-(Trifluoromethyl)picolinic acid
Formula:
C₇H₄F₃NO₂ Peso molecolare: 191,1100 g/mol |
MDL Number:
MFCD07375386 CAS Number: 87407-12-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...