Stampa…

(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: (R)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthalene, (R)-(+)-TolBINAP

T3152-1G T3152-5G
TCIAT3152-1GEA 51 EUR
TCIAT3152-1G TCIAT3152-5G
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98.0% (by HPLC)
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Formula: C₄₈H₄₀P₂
Peso molecolare: 678,78 g/mol
Punto di fusione: 258°C
MDL Number: MFCD01311709
CAS Number: 99646-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cdb1862eca03966ee79c61c8f3bed319b294d25&vid=0cdb1862eca03966ee79c61c8f3bed319b294d25&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR