Stampa…

Tetrakis(4-ethynylphenyl)methane ≥98.0% (by GC)

Fornitore: TCI EUROPE

T3151-100MG T3151-1G
TCIAT3151-100MGEA 120 EUR
TCIAT3151-100MG TCIAT3151-1G
Tetrakis(4-ethynylphenyl)methane ≥98.0% (by GC)
Tetrakis(4-ethynylphenyl)methane
Formula: C₃₃H₂₀
Peso molecolare: 416,52 g/mol
Punto di fusione: 300°C
MDL Number: MFCD24873189
CAS Number: 177991-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1054f6278fbded420864d03f0fbe469e5b49358c&vid=1054f6278fbded420864d03f0fbe469e5b49358c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR