2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(2-Pyrenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(pyren-2-yl)-1,3,2-dioxaborolane, 2-Pyreneboronic acid Pinacol ester
T3150-1G
T3150-5G
TCIAT3150-1GEA
285
EUR
TCIAT3150-1G
TCIAT3150-5G
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene ≥98.0% (by GC)
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene
Formula:
C₂₂H₂₁BO₂ Peso molecolare: 328,212 g/mol |
MDL Number:
MFCD11977261 CAS Number: 853377-11-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...