Stampa…

Tin(II) 2-ethylhexanoate ≥85.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Ethylcaproic acid tin (II) salt, Tin bis(2-ethylhexanoate), Stannous 2-ethylhexanoate, 2-Ethylhexanoic acid tin (II) salt

T3149-100G T3149-500G
TCIAT3149-100GEA 27 EUR
TCIAT3149-100G TCIAT3149-500G
Tin(II) 2-ethylhexanoate ≥85.0% (by titrimetric analysis)
Tin(II) 2-ethylhexanoate
Formula: C₁₆H₃₀O₄Sn
Peso molecolare: 405.12 g/mol
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb4e87c02a2fc4bf8a50e7596855317ac7283b45&vid=fb4e87c02a2fc4bf8a50e7596855317ac7283b45&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (Iodometric Titration) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR