Stampa…

N-(Trifluoromethylthio)phthalimide ≥98.0% (by GC)

Fornitore: TCI
Attenzione

T3143-1G T3143-5G
TCIAT3143-1GEA 168 EUR
TCIAT3143-1G TCIAT3143-5G
N-(Trifluoromethylthio)phthalimide ≥98.0% (by GC)
N-(Trifluoromethylthio)phthalimide
Formula: C₉H₄F₃NO₂S
Peso molecolare: 247.19 g/mol
MDL Number: MFCD18449627
CAS Number: 719-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c84f01dfe78ab8fab8004a29410f7a1b81ec7405&vid=c84f01dfe78ab8fab8004a29410f7a1b81ec7405&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR