Stampa…

1,3,5-Tris(2-propynyloxy)benzene ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 1,3,5-Tris(propargyloxy)benzene

T3135-200MG T3135-1G
TCIAT3135-200MGEA 158 EUR
TCIAT3135-200MG TCIAT3135-1G
1,3,5-Tris(2-propynyloxy)benzene ≥98.0% (by titrimetric analysis)
1,3,5-Tris(2-propynyloxy)benzene
Formula: C₁₅H₁₂O₃
Peso molecolare: 240,26 g/mol
MDL Number: MFCD28384155
CAS Number: 114233-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81e9c65ea633e01ce5c7de17df137e786a32f0ef&vid=81e9c65ea633e01ce5c7de17df137e786a32f0ef&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR